Dextrin, precipitated by alcohol
Glass bottle for solids, 100 g
Leverandør: Thermo Fisher Scientific
Dextrin, precipitated by alcohol
Katalog # A15717.22
Leverandør: Thermo Fisher Scientific
CAS nummer: 9004-53-9
Gå til:
Product Details & Documents
CAS No.: 9004-53-9 Merck Reference: 14,2953 MDL No.: MFCD00081554
Synonymer: Dextrin
- CAS nummer:9004-53-9
- Formel:(C₆H₁₂O₆)n
- MDL nummer:MFCD00081554
- Molecular Weight:180.16 g/mol
- Merck Index:14,2953
- Tæthed:1,45 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:232-675-4
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:100 g



