Olie rød O
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
CAS No.: 1320-06-5 Molecular Formula: C26H24N4O Formula Weight: 408.51 Melting Point: ca 166° dec. Beilstein Reference: 3491382 MDL No.: MFCD00003898
Synonyms: CI 26125
- CAS Number:1320-06-5
- Formula:C₂₆H₂₄N₄O
- MDL Number:MFCD00003898
- Molecular Weight:408.51 g/mol
- Melting Pt:∼ 166 °C (dec.)
- Storage Temperature:Stue temperatur
- EINECS:215-295-3
Documents
Specifications
Specifications
- Pakning:Glass bottle for solids
- Pakn:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



