Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Kaliumsorbat ≥99%

Kaliumsorbat ≥99%

Glass bottle for solids, 1.000 g
Katalog # A12844.0B
Irriterende
Leverandør:   Thermo Fisher Scientific
Kaliumsorbat ≥99%
Katalog # A12844.0B
CAS nummer:  24634-61-5
Kaliumsorbat ≥99%
Kaliumsorbat ≥99%

Glass bottle for solids, 1.000 g

CAS nummer:  24634-61-5
Price
1.200,00 kr
Log ind for tilgængelighed
1 * 1.000 g
Product Details & Documents

Purity: 99% CAS No.: 24634-61-5 Molecular Formula: C6H7KO2 Formula Weight: 150.22 pH: 8,3 Melting Point: 300° MDL No.: MFCD00016546

Synonymer:  Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate
  • CAS nummer:24634-61-5
  • Formel:CH₃CH=CHCH=CHCOOK
  • MDL nummer:MFCD00016546
  • Molecular Weight:150.22 g/mol
  • Merck Index:14,7671
  • Smeltende Pt:>300 °C
  • Tæthed:1,361
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:246-376-1
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    1.000 g

Specifikationer

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.