Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

3'-Chlorpropiophenon ≥98%

Glass bottle for solids, 500 g
Katalog # A12542.36
Leverandør:   Thermo Fisher Scientific
3'-Chlorpropiophenon ≥98%
Katalog # A12542.36
CAS nummer:  34841-35-5
3'-Chlorpropiophenon ≥98%
3'-Chlorpropiophenon ≥98%

Glass bottle for solids, 500 g

CAS nummer:  34841-35-5
Price
4.700,00 kr
Log ind for tilgængelighed
1 * 500 g
Product Details & Documents

Purity: 98% CAS No.: 34841-35-5 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 45-47° Boiling Point: 123-124°/13mm MDL No.: MFCD00009925

Synonymer:  m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one
  • CAS nummer:34841-35-5
  • Formel:C₉H₉ClO
  • MDL nummer:MFCD00009925
  • Molecular Weight:168.62 g/mol
  • Kogende Pt:123…124 °C (13 mmHg)
  • Smeltende Pt:45…47 °C
  • Flash Pt:>110 °C (230 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:252-242-3
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    500 g

Specifikationer