Orders LinkContactUsLinkComponent
3'-Chlorpropiophenon ≥98%
3'-Chlorpropiophenon ≥98%
Glass bottle for solids, 100 g
  A12542.22
3'-Chlorpropiophenon ≥98%
  A12542.22
 :  34841-35-5
3'-Chlorpropiophenon ≥98%
3'-Chlorpropiophenon ≥98%
Glass bottle for solids, 100 g
 :  34841-35-5
1.200,00 kr
1 * 100 g
Purity: 98% CAS No.: 34841-35-5 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 45-47° Boiling Point: 123-124°/13mm MDL No.: MFCD00009925
 :  m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one
  •  :34841-35-5
  •  :C₉H₉ClO
  •  :MFCD00009925
  •  :168.62 g/mol
  •  :123…124 °C (13 mmHg)
  •  :45…47 °C
  •  :>110 °C (230 °F)
  •  :Stue temperatur
  •  :252-242-3
  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 g