3'-Chlorpropiophenon ≥98%
Glass bottle for solids, 100 g
Purity: 98% CAS No.: 34841-35-5 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 45-47° Boiling Point: 123-124°/13mm MDL No.: MFCD00009925
: m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one
- :34841-35-5
- :C₉H₉ClO
- :MFCD00009925
- :168.62 g/mol
- :123…124 °C (13 mmHg)
- :45…47 °C
- :>110 °C (230 °F)
- :Stue temperatur
- :252-242-3
Documents
- Pakning:Glass bottle for solids
- Pakn:100 g