Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

3'-Chlorpropiophenon ≥98%

Glass bottle for solids, 25 g
Katalog # A12542.14
Leverandør:   Thermo Fisher Scientific
3'-Chlorpropiophenon ≥98%
Katalog # A12542.14
CAS nummer:  34841-35-5
3'-Chlorpropiophenon ≥98%
3'-Chlorpropiophenon ≥98%

Glass bottle for solids, 25 g

CAS nummer:  34841-35-5
Price
458,00 kr
Log ind for tilgængelighed
1 * 25 g
Product Details & Documents

Purity: 98% CAS No.: 34841-35-5 Molecular Formula: C9H9ClO Formula Weight: 168.62 Flash Point: 110°(230°F) Melting Point: 45-47° Boiling Point: 123-124°/13mm MDL No.: MFCD00009925

Synonymer:  m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one
  • CAS nummer:34841-35-5
  • Formel:C₉H₉ClO
  • MDL nummer:MFCD00009925
  • Molecular Weight:168.62 g/mol
  • Kogende Pt:123…124 °C (13 mmHg)
  • Smeltende Pt:45…47 °C
  • Flash Pt:>110 °C (230 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:252-242-3
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    25 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.