Cyclohexanone-2,4-dinitrophenylhydrazone 99%
25 g
Leverandør: Thermo Fisher Scientific
Cyclohexanone-2,4-dinitrophenylhydrazone 99%
Katalog # A12203.14
Leverandør: Thermo Fisher Scientific
CAS nummer: 1589-62-4
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 1589-62-4 Molecular Formula: C12H14N4O4 Formula Weight: 278.27 Melting Point: 158-160° MDL No.: MFCD00001658
- CAS nummer:1589-62-4
- Formel:C₁₂H₁₄N₄O₄
- MDL nummer:MFCD00001658
- Molecular Weight:278.27 g/mol
- Smeltende Pt:158…160 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:216-458-1
Specifikationer
Specifikationer
- Pakn:25 g



