Syringes for CTC analytics systems
Fixed needle, 26, 10 µl, 50 mm, 0,47 mm, koniske, 10F-CTC-5/0.47C
554-1442
* gastæt
- Udv.Ø:0,47 mm
- Måler:26
- Model:10F-CTC-5/0.47C
- Spidstype:koniske
- Volumen:10 µl
- Beskrivelse:Fixed needle
- Længde:50 mm
- Seq:23