Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Capillaries for LC systems

Capillaries for LC systems

Restriction capillary, I.Ø 0,17 mm
Katalog # HEWLG1312-87301
Leverandør:   AGILENT HEWLETT PACKARD
Capillaries for LC systems
Katalog # HEWLG1312-87301
Capillaries for LC systems
Capillaries for LC systems

Restriction capillary, I.Ø 0,17 mm

Price
1.280,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Stainless steel capillaries are made of flexible stainless steel (0,6 mm OD) with ¹/₁₆" OD tubing at both ends to accept standard fittings.

Pre-swaged fittings are assembled according to Swagelok specifications.

Specifikationer
  • Beskrivelse:
    Restriction capillary, I.Ø 0,17 mm
  • Seq:
    109