Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Acenaphthen-D10 (99% D)

Acenaphthen-D10 (99% D)

Glass bottle for solids, 100 mg
Katalog # CAMBDLM-108-0.1
Miljø
Leverandør:   CAMBRIDGE ISOTOPE LABS
Acenaphthen-D10 (99% D)
Katalog # CAMBDLM-108-0.1
Leverandør:  CAMBRIDGE ISOTOPE LABS
CAS nummer:  15067-26-2
Acenaphthen-D10 (99% D)

Glass bottle for solids, 100 mg

CAS nummer:  15067-26-2
Price
Spørg for pris
Log ind for tilgængelighed
1 * 100 mg
Product Details & Documents
Synonymer:  Acenaphthene-[²H₁₀]
  • CAS nummer:15067-26-2
  • Formel:C₁₂D₁₀
  • Molecular Weight:164.13 g/mol
  • Opbevaringstemperatur:Stue temperatur
  • FN:3077
  • ADR:9,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    100 mg

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.