Universal couplings, BOLA
internal thread M 6, female, UNF ¼" 28 G, outer thread, PTFE
Katalog # BOHLF770-08
Leverandør: Bohlender
Gå til:
Product Details & Documents
PTFE, white.
- Temperature resistance: from –200 to +250 °C
- Universal chemical resistance
- FDA conform
Coupling for transition from M6 thread to UNF ¼" 28 G thread. The M6 thread is marked by a groove.
Specifikationer
- Beskrivelse:internal thread M 6
- Materiale:PTFE
- Gevind:female, UNF ¼" 28 G, outer thread