Sigmacote® siliconizing reagent
25 mL
Katalog # SL2-25ML
Leverandør: SIGMA ALDRICH MICROSCOPY
Gå til:
Product Details & Documents
Siliconizing reagent for glass and other surfaces. Sigmacote® is a solution of a chlorinated organopolysiloxane in heptane that readily forms a covalent, microscopically thin film on glass.
- ADR:8(3),II
- :M-Clarity™ quality level = MQ200
- Flash Pt:–4 °C
- Opbevaringstemperatur:Køleskab
- FN:2924
Documents
Specifikationer
Specifikationer
- Pakn:25 mL