Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Sigmacote® siliconizing reagent

Sigmacote® siliconizing reagent

100 mL
Katalog # SL2-100ML
Brandfarlig
Irriterende
Sundhed
Miljø
Leverandør:   SIGMA ALDRICH MICROSCOPY
Sigmacote® siliconizing reagent
Katalog # SL2-100ML
CAS nummer:  
Sigmacote® siliconizing reagent

100 mL

CAS nummer:  
Price
1.800,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents

Siliconizing reagent for glass and other surfaces. Sigmacote® is a solution of a chlorinated organopolysiloxane in heptane that readily forms a covalent, microscopically thin film on glass.

  • ADR:8(3),II
  •  :M-Clarity™ quality level = MQ200
  • Flash Pt:–4 °C
  • Opbevaringstemperatur:Køleskab
  • FN:2924
Specifikationer

Specifikationer

  • Pakn:
    100 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.