Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Saffron, Sigma-Aldrich®

Saffron, Sigma-Aldrich®

Glass bottle for solids, 5 g
Katalog # S8381-5G
Leverandør:   SIGMA ALDRICH MICROSCOPY
Saffron, Sigma-Aldrich®
Katalog # S8381-5G
CAS nummer:  84604-17-1
Saffron, Sigma-Aldrich®
Saffron, Sigma-Aldrich®

Glass bottle for solids, 5 g

CAS nummer:  84604-17-1
Price
3.080,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  C.I. 75100
  • CAS nummer:84604-17-1
  • Molecular Weight:976.96 g/mol
  •  :M-Clarity™ quality level = MQ200
  • EINECS:283-295-0
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.