Methylcyclohexan 99%, Reagent Grade
Glass bottle, 1 L
Leverandør: Honeywell Chemicals
Methylcyclohexan 99%, Reagent Grade
Katalog # M37889-1L
Leverandør: Honeywell Chemicals
CAS nummer: 108-87-2
Gå til:
Product Details & Documents
- ≥95% for general laboratory use.
- CAS nummer:108-87-2
- Formel:C₆H₁₁CH₃
- MDL nummer:MFCD00001497
- Molecular Weight:98.19 g/mol
- Merck Index:13,06074
- Kogende Pt:6,7 %(V)
- Smeltende Pt:–126 °C
- Smeltende Pt:-126 °C
- Tæthed:0,769 g/cm³ (20 °C)
- Flash Pt:–4 °C
- Flash Pt:-4 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-624-3
- FN:2296
- ADR:3,II
Documents
sds_M37889
SDS
Specifikationer
Specifikationstestresultater
- Identity (IR):complying
- Assay (GC):Min. 98.5 %
- Vapor Pressure:83.2 mmHg ( 37.7 °C)
- Refractive Index:n20/D 1.422 (lit.)
- Vapor Density:3.4 (vs air)
- Autoignition Temperature:545 °F
Specifikationer
- Pakning:Glass bottle
- Pakn:1 L
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



