Orders LinkContactUsLinkComponent
Microtube refill system for micro-racks, ratiolab®
Microtube refill system for micro-racks, ratiolab®
refill with 96 microtubes, non-coded, 0,65 ml, 50 pcs/pk
Katalog # RATI8200025
Leverandør:   RATIOLAB
Microtube refill system for micro-racks, ratiolab®
Katalog # RATI8200025
Leverandør:  RATIOLAB
Microtube refill system for micro-racks, ratiolab®
Microtube refill system for micro-racks, ratiolab®
refill with 96 microtubes, non-coded, 0,65 ml, 50 pcs/pk
Price
2.290,00 kr
Log ind for tilgængelighed
1 * 4.800 Items
Product Details & Documents
Plastic rack with cardboard box.
  • Available with coded or non-coded Micro-Tubes
  • For the very efficient, hygienic, and fast loading of Micro-Racks
  • Convenient handling and reduced waste
  • Pressure-tested tightness of the Micro-Tubes
The Micro-Tubes are placed on an 8×12 tray and packed in an environmentally friendly cardboard box. The loaded tray can easily be moved from the cardboard box into the empty Micro-Rack.
Specifikationer
  • Pakket:
    50 pcs/pk
  • Kapacitet:
    0,65 ml
  • Beskrivelse:
    refill with 96 microtubes, non-coded