Microtube racks
Mikrorørstativ, fyldt, kodet, gennemsigtig, 96, 1,2 ml, Round bottom
Katalog # RATI8200004
Leverandør: RATIOLAB
Gå til:
Product Details & Documents
Racks, made of PP, translucent, for micro tubes, feature a highly practical system for processing small volumes and a stable robust system having a dust tight lid and also available as empty racks or equipped with coded or non-coded.
- Side edges labeled with a barcode
- Stackable to use with stacker instruments
- Sloping edge helps the positioning of the refill unit
- Temperature resistant to −80 °C
Specifikationer
- Til rør:1,2 ml
- Form:Round bottom
- Ant. huller:96
- Farve:gennemsigtig
- Beskrivelse:Mikrorørstativ, fyldt, kodet
- Seq:03