Orders LinkContactUsLinkComponent
Thermal transfer printer labels, type B-7606
Printer labels, B-7606, Papir, Permanent akryl, Thermal transfer, hvid, BBP12, 101×38 mm, 11520 labels/box
Katalog # BRDY236734
Leverandør:   Brady
Thermal transfer printer labels, type B-7606
Katalog # BRDY236734
Leverandør:  Brady
Thermal transfer printer labels, type B-7606
Thermal transfer printer labels, type B-7606
Printer labels, B-7606, Papir, Permanent akryl, Thermal transfer, hvid, BBP12, 101×38 mm, 11520 labels/box
Price
55.780,00 kr
Log ind for tilgængelighed
1 * 12 ROL
Product Details & Documents
Printable, paper labels for thermal transfer printers.
Specifikationer
  • Printer:
    BBP12
  • Printing method:
    Thermal transfer
  • Model:
    B-7606
  • Farve:
    hvid
  • Beskrivelse:
    Printer labels
  • Dimensioner:
    101×38 mm
  • Materiale:
    Papir
  • Seq:
    05
  • Pakket:
    11520 labels/box
  • Klæbestof:
    Permanent akryl