Interchangeable blocks for block heaters, digital, SB1
Interchangeable block, 50 ml centrifuge tubes, 5
Katalog # JENC357-136
Leverandør: JENCONS SCIENTIFIC
Interchangeable blocks for block heaters, digital, SB1
Katalog # JENC357-136
Leverandør: JENCONS SCIENTIFIC
Gå til:
Product Details & Documents
Interchangeable blocks are made of high grade, non porous, anodised aluminium. Except the blocks used for plates and slides, all other blocks include positions for a block lifter and calibration thermometer.
Specifikationer
- Ant. huller:5
- Beskrivelse:Interchangeable block
- Seq:15
- Til:50 ml centrifuge tubes