Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Accessories for TSK-Gel® amide-80 columns

Accessories for TSK-Gel® amide-80 columns

Guard cartridge holder for 2 mm I-Ø guard cartridges, 2,00 mm, 15 mm
Katalog # TOYO0019308
Leverandør:   TOSOH Bioscience
Accessories for TSK-Gel® amide-80 columns
Katalog # TOYO0019308
Leverandør:  TOSOH Bioscience
Accessories for TSK-Gel® amide-80 columns
Accessories for TSK-Gel® amide-80 columns

Guard cartridge holder for 2 mm I-Ø guard cartridges, 2,00 mm, 15 mm

Price
3.540,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Specifikationer
  • Ø indv.:
    2,00 mm
  • Beskrivelse:
    Guard cartridge holder for 2 mm I-Ø guard cartridges
  • Længde:
    15 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.