Orders LinkContactUsLinkComponent
General purpose thermometers
0…+50 °C, 1,0 °C, Rød sprit, 7,0 - 8,0 mm
  HECH43201008
General purpose thermometers
  HECH43201008
General purpose thermometers
General purpose thermometers
0…+50 °C, 1,0 °C, Rød sprit, 7,0 - 8,0 mm
181,00 kr
1 * 1 Items
General purpose thermometers with red filling.
  • Ø:
    7,0 - 8,0 mm
  • Seq:
    01
  • Inddeling:
    1,0 °C
  • Påfyldning:
    Rød sprit
  • Område:
    0…+50 °C