Orders LinkContactUsLinkComponent
Wooden applicators
One pointed end applicators for cotton wool, Hårdttræ, 200 mm
Katalog # HECH44301020
Wooden applicators
Katalog # HECH44301020
Wooden applicators
Wooden applicators
One pointed end applicators for cotton wool, Hårdttræ, 200 mm
Price
32,30 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
Wooden applicators of hardwood in bundles of 50 or 100 pieces.
  • Unpointed (blunt), one end pointed and cotton ended variants are available
 : Unpointed (blunt), one end pointed and cotton ended variants.
 : *Produktet/produkterne på denne side er uden for anvendelsesområdet for EU -direktiv 2019/904
Specifikationer
  • Beskrivelse:
    One pointed end applicators for cotton wool
  • Længde:
    200 mm
  • Materiale:
    Hårdttræ
  • Seq:
    04