Orders LinkContactUsLinkComponent
Powder funnel, short stem
160 mm, 35 mm, 24 mm
Katalog # HECH44001052
Powder funnel, short stem
Katalog # HECH44001052
Powder funnel, short stem
Powder funnel, short stem
160 mm, 35 mm, 24 mm
Price
kr534.00
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Soda-Kalkglas.
Specifikationer
  • Tragt udv. Ø:
    160 mm
  • Seq:
    04
  • Stilk udv. Ø:
    35 mm
  • Stilklængde:
    24 mm