Funnels with short stem
80 mm
Katalog # HECH49050006
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Funnels, made of HDPE, translucent. With smooth internal walls. Robust and strong. Multipurpose funnel for a wide variety of uses.
- Good chemical resistance
- Suspension eye for easy storage
- Easy to clean
Specifikationer
- Tragt udv. Ø:80 mm