Flasks, round bottom, with standard ground joints
250 ml, 29/32 NS
Katalog # HECH41913018
Leverandør: GLASWARENFABRIK KARL HECHT
Flasks, round bottom, with standard ground joints
Katalog # HECH41913018
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.
Specifikationer
- Kappestr.:29/32 NS
- Kapacitet:250 ml