Orders LinkContactUsLinkComponent
Flasks, round bottom, with standard ground joints
Flasks, round bottom, with standard ground joints
100 ml, 29/32 NS
Katalog # HECH41913012
Flasks, round bottom, with standard ground joints
Katalog # HECH41913012
Flasks, round bottom, with standard ground joints
Flasks, round bottom, with standard ground joints
100 ml, 29/32 NS
Price
159,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.
Specifikationer
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    100 ml