Flasks, round bottom, with standard ground joints
50 ml, 19/26 NS
Catalog # HECH41913010
Supplier: GLASWARENFABRIK KARL HECHT
Flasks, round bottom, with standard ground joints
Catalog # HECH41913010
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.
Specifications
- Kappestr.:19/26 NS
- Kapacitet:50 ml