Orders LinkContactUsLinkComponent
Flasks, round bottom, with standard ground joints
Flasks, round bottom, with standard ground joints
50 ml, 19/26 NS
Catalog # HECH41913010
Flasks, round bottom, with standard ground joints
Catalog # HECH41913010
Flasks, round bottom, with standard ground joints
Flasks, round bottom, with standard ground joints
50 ml, 19/26 NS
Price
144,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One centre neck, useful for distillation and extraction.
Specifications
  • Kappestr.:
    19/26 NS
  • Kapacitet:
    50 ml