Rubber Stoppers
55,0 mm, 64,5 mm, 75,5 mm
Catalog # HECH41590059
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
DIN 12871, made of natural rubber, hardness 40 Shore A.
Documents
HECH41590041_TDS_EN
Datasheet
Specifications
- Ø bund:64,5 mm
- Ø top:75,5 mm
- Højde:55,0 mm
- Seq:15