Orders LinkContactUsLinkComponent
Sleeves for ground joints
Sleeves for ground joints
29/32
Catalog # HECH42388029
Sleeves for ground joints
Catalog # HECH42388029
Sleeves for ground joints
Sleeves for ground joints
29/32
Price
268,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Disposable sleeves for ground joints and stoppers, prevent the ground joints from sticking together.
  • Made of PTFE
  • Self-lubricating
  • Outstanding chemical resistance at temperatures
Specifications
  • Til NS:
    29/32
  • Seq:
    04