Sleeves for ground joints
29/32
Catalog # HECH42388029
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Disposable sleeves for ground joints and stoppers, prevent the ground joints from sticking together.
- Made of PTFE
- Self-lubricating
- Outstanding chemical resistance at temperatures
Specifications
- Til NS:29/32
- Seq:04