Desiccator plates
Porcelain, 200 mm
Catalog # HECH41175020
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Porcelain plates for desiccators with many small holes, without base.
Specifications
- Ø:200 mm
- Model:Porcelain
Porcelain plates for desiccators with many small holes, without base.