Orders LinkContactUsLinkComponent
Desiccator plates
Desiccator plates
Porcelain, 200 mm
Catalog # HECH41175020
Desiccator plates
Catalog # HECH41175020
Desiccator plates
Desiccator plates
Porcelain, 200 mm
Price
529,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Porcelain plates for desiccators with many small holes, without base.
Specifications
  • Ø:
    200 mm
  • Model:
    Porcelain