Orders LinkContactUsLinkComponent
Desiccator plates
Desiccator plates
Porcelain, 150 mm
Katalog # HECH41175015
Desiccator plates
Katalog # HECH41175015
Desiccator plates
Desiccator plates
Porcelain, 150 mm
Price
283,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Porcelain plates for desiccators with many small holes, without base.
Specifikationer
  • Ø:
    150 mm
  • Model:
    Porcelain