Desiccator plates
Porcelain, 100 mm
Katalog # HECH41175010
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Porcelain plates for desiccators with many small holes, without base.
Specifikationer
- Ø:100 mm
- Model:Porcelain