Pressure bulbs
Rubber bulbs, single, with valve and tubing
Katalog # HECH40850001
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
simple: with 2 valves – made of soft PVC (cadmium-free)
double: with valve - made of soft PVC (cadmium-free), ball in the net made of latex
Specifikationer
- Beskrivelse:Rubber bulbs, single, with valve and tubing
- Seq:01