Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
2000 ml, 29/32, 0,60 ± ml
Katalog # HECH92209800810
Volumetric flasks, class A, blue markings
Katalog # HECH92209800810
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
2000 ml, 29/32, 0,60 ± ml
Price
805,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in blue
  • Conformity certified (DE-M marking)
Certifications: DIN EN ISO 1042
Documents
Specifikationer
  • NS:
    29/32
  • Kapacitet:
    2000 ml
  • Seq:
    12
  • Tolerance:
    0,60 ± ml