Volumetric flasks, class A, blue markings
2000 ml, 29/32, 0,60 ± ml
Katalog # HECH92209800810
Leverandør: GLASWARENFABRIK KARL HECHT
Volumetric flasks, class A, blue markings
Katalog # HECH92209800810
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Calibrated to contain (TC, In)
- Marks and inscriptions in blue
- Conformity certified (DE-M marking)
Certifications: DIN EN ISO 1042
Documents
Certifikater
Søg efter partinummerSpecifikationer
- NS:29/32
- Kapacitet:2000 ml
- Seq:12
- Tolerance:0,60 ± ml