Orders LinkContactUsLinkComponent
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
200 ml, 14/23, 0,15 ± ml
Katalog # HECH92209800509
Volumetric flasks, class A, blue markings
Katalog # HECH92209800509
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings
200 ml, 14/23, 0,15 ± ml
Price
2.320,00 kr
Log ind for tilgængelighed
1 * 10 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Calibrated to contain (TC, In)
  • Marks and inscriptions in blue
  • Conformity certified (DE-M marking)
Certifications: DIN EN ISO 1042
Documents
Specifikationer
  • NS:
    14/23
  • Kapacitet:
    200 ml
  • Seq:
    08
  • Tolerance:
    0,15 ± ml