Volumetric flasks, class A, blue markings
200 ml, 14/23, 0,15 ± ml
Katalog # HECH92209800509
Leverandør: GLASWARENFABRIK KARL HECHT
Volumetric flasks, class A, blue markings
Katalog # HECH92209800509
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Borosilicate glass 3.3.
- Calibrated to contain (TC, In)
- Marks and inscriptions in blue
- Conformity certified (DE-M marking)
Certifications: DIN EN ISO 1042
Documents
Certifikater
Søg efter partinummerSpecifikationer
- NS:14/23
- Kapacitet:200 ml
- Seq:08
- Tolerance:0,15 ± ml