Clips for conical ground joints
orange, 34/35
HECH42386034
Clips made of POM for conical ground joints.
- Material does not scratch glass surfaces
- Resistant to temperatures up to +150 °C
- Til NS:34/35
- Farve:orange
- Seq:07
Clips made of POM for conical ground joints.