Clips for conical ground joints
lilla, 12/21
Katalog # HECH42386012
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Clips made of POM for conical ground joints.
- Material does not scratch glass surfaces
- Resistant to temperatures up to +150 °C
Specifikationer
- Til NS:12/21
- Farve:lilla
- Seq:02