Clips for conical ground joints
grøn, 10/19
Katalog # HECH42386010
Leverandør: GLASWARENFABRIK KARL HECHT
Gå til:
Product Details & Documents
Clips made of POM for conical ground joints.
- Material does not scratch glass surfaces
- Resistant to temperatures up to +150 °C
Specifikationer
- Til NS:10/19
- Farve:grøn
- Seq:01