Orders LinkContactUsLinkComponent
Screw top vials
Screw top vials
Vials, 2,00 ml, 12×32 mm, klar, No, 8-425
Katalog # SIAL27078
Leverandør:   Merck
Screw top vials
Katalog # SIAL27078
Leverandør:  Merck
Screw top vials
Vials, 2,00 ml, 12×32 mm, klar, No, 8-425
Price
kr294.00
Log ind for tilgængelighed
1 * 100 Items
This product has been blocked by your organization. Please contact your purchasing department for more information
Product Details & Documents
Specifikationer
  • Kasket inkluderet:
    No
  • Cap størrelse:
    8-425
  • Diameter×H:
    12×32 mm
  • Kapacitet:
    2,00 ml
  • Farve:
    klar
  • Beskrivelse:
    Vials
  • Seq:
    08
  • Cat. No.:
    SIAL27078