Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Jod ≥99.8%, solid ACS, Fluka™

Glass bottle, 100 g
Katalog # 207772-100G
Sundhed
Irriterende
Leverandør:   Honeywell Chemicals
Jod ≥99.8%, solid ACS, Fluka™
Katalog # 207772-100G
Leverandør:  Honeywell Chemicals
CAS nummer:  7553-56-2
Jod ≥99.8%, solid ACS, Fluka™

Glass bottle, 100 g

CAS nummer:  7553-56-2
Price
883,00 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents

Honeywell ACS solvents for general laboratory use are:

  • A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • Suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested meet ACS general use specifications
  • CAS nummer:7553-56-2
  • Formel:I₂
  • MDL nummer:MFCD00011355
  • Molecular Weight:253.81 g/mol
  • Smeltende Pt:113,5 °C
  • Tæthed:4,930 g/cm³ (20 °C)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:231-442-4
  • FN:3495
  • ADR:8,III
Specifikationer

Specifikationstestresultater

  • Assay:
    Min. 99.8 %
  • Non-volatile matter:
    Max. 0.01 %
  • Bromine and chlorine (as chloride):
    Max. 0.005 %
  • Form:
    Solid
  • Vapor Pressure:
    0.31 mmHg ( 25 °C)
  • Resistivity:
    1.3E15 μΩ-cm

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    100 g

Specifikationer