n-Hexan ≥99%, Reagent Grade
Glass bottle, 3.000 mL
Leverandør: Honeywell Chemicals
n-Hexan ≥99%, Reagent Grade
Katalog # 139386-6X500ML
Leverandør: Honeywell Chemicals
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
- ≥95% for general laboratory use.
Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:66…69 °C(1,013 hPa)
- Smeltende Pt:–95 °C
- Tæthed:0,660…0.668 g/cm³ (20 °C)
- Flash Pt:–26 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-777-6
- FN:1208
- ADR:3,II
Documents
Specifikationer
Specifikationstestresultater
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.373 - 1.377
- Vapor Pressure:256 mmHg ( 37.7 °C)
- Vapor Density:~3 (vs air)
- Refractive Index:n20/D 1.375 (lit.)
- Explosion Limit:7.7 %
Specifikationer
- Pakning:Glass bottle
- Pakn:3.000 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



