n-Hexan ≥99%, Reagent Grade
Glass bottle, 500 mL
Leverandør: Honeywell Chemicals
n-Hexan ≥99%, Reagent Grade
Katalog # 139386-500ML
Leverandør: Honeywell Chemicals
CAS nummer: 110-54-3
Gå til:
Product Details & Documents
- ≥95% for general laboratory use.
Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502
Synonymer: Hexyl hydride, Hexanen, Hexane
- CAS nummer:110-54-3
- Formel:H₃C(CH₂)₄CH₃
- MDL nummer:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Kogende Pt:66…69 °C(1,013 hPa)
- Smeltende Pt:–95 °C
- Tæthed:0,660…0.668 g/cm³ (20 °C)
- Flash Pt:–26 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-777-6
- FN:1208
- ADR:3,II
Specifikationer
Specifikationstestresultater
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.373 - 1.377
- Vapor Pressure:256 mmHg ( 37.7 °C)
- Vapor Density:~3 (vs air)
- Refractive Index:n20/D 1.375 (lit.)
- Explosion Limit:7.7 %
Specifikationer
- Pakning:Glass bottle
- Pakn:500 mL



