n-Hexan ≥99%, Reagent Grade
Glass bottle, 2,5 L
Supplier: Honeywell Chemicals
Jump to:
Product Details & Documents
- ≥95% for general laboratory use.
Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:66…69 °C(1,013 hPa)
- Melting Pt:–95 °C
- Density:0,660…0.668 g/cm³ (20 °C)
- Flash Pt:–26 °C
- Storage Temperature:Stue temperatur
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Documents
Specifications
Specification Test Results
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Refractive index (n 20/D):1.373 - 1.377
- Vapor Pressure:256 mmHg ( 37.7 °C)
- Vapor Density:~3 (vs air)
- Refractive Index:n20/D 1.375 (lit.)
- Explosion Limit:7.7 %
Specifications
- Pakning:Glass bottle
- Pakn:2,5 L



