Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
n-Hexan ≥99%, Reagent Grade

n-Hexan ≥99%, Reagent Grade

Glass bottle, 100 mL
Katalog # 139386-100ML
Irriterende
Sundhed
Brandfarlig
Leverandør:   Honeywell Chemicals
n-Hexan ≥99%, Reagent Grade
Katalog # 139386-100ML
Leverandør:  Honeywell Chemicals
CAS nummer:  110-54-3
n-Hexan ≥99%, Reagent Grade
n-Hexan ≥99%, Reagent Grade

Glass bottle, 100 mL

CAS nummer:  110-54-3
Price
203,00 kr
Log ind for tilgængelighed
1 * 100 mL
Product Details & Documents
  • ≥95% for general laboratory use.

Beilstein Number: 1730733
RTECS Number: MN9275000
UNSPSC Code: 12191502

Synonymer:  Hexyl hydride, Hexanen, Hexane
  • CAS nummer:110-54-3
  • Formel:H₃C(CH₂)₄CH₃
  • MDL nummer:MFCD00009520
  • Molecular Weight:86.18 g/mol
  • Merck Index:13,04712
  • Kogende Pt:66…69 °C(1,013 hPa)
  • Smeltende Pt:–95 °C
  • Tæthed:0,660…0.668 g/cm³ (20 °C)
  • Flash Pt:–26 °C
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:203-777-6
  • FN:1208
  • ADR:3,II

Documents

Specifikationer

Specifikationstestresultater

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.0 %
  • Refractive index (n 20/D):
    1.373 - 1.377
  • Vapor Pressure:
    256 mmHg ( 37.7 °C)
  • Vapor Density:
    ~3 (vs air)
  • Refractive Index:
    n20/D 1.375 (lit.)
  • Explosion Limit:
    7.7 %

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    100 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.