Objektglas med fordybning
Objektglas med reaktionsbrønd, sort, 1, 6 mm
Katalog # MARI1216540
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
- Soda-lime glass with epoxy resin colour masks
- High resistance against solvents
Documents
Specifikationer
- Ant. brønde:1
- Farve:sort
- Beskrivelse:Objektglas med reaktionsbrønd
- Seq:01
- Brønd Ø:6 mm