Objektglas med fordybning
Objektglas med reaktionsbrønde, hvid, 6, 8 mm
Katalog # MARI1215132
Leverandør: MARIENFELD
Gå til:
Product Details & Documents
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
- Soda-lime glass with epoxy resin colour masks
- High resistance against solvents
Documents
Specifikationer
- Ant. brønde:6
- Farve:hvid
- Beskrivelse:Objektglas med reaktionsbrønde
- Seq:14
- Brønd Ø:8 mm