Objektglas med fordybning
Objektglas med reaktionsbrønde, hvid, 10, 5 mm
Catalog # MARI1216652
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Cavity slides for diagnosis with well wettable reaction wells and frosted marking area of 20 mm at one end.
- Soda-lime glass with epoxy resin colour masks
- High resistance against solvents
Documents
Specifications
- Ant. brønde:10
- Farve:hvid
- Beskrivelse:Objektglas med reaktionsbrønde
- Seq:20
- Brønd Ø:5 mm