Tilbehør til printer og vægte
Product Details & Documents
The SF40A impact printer and the STP103 thermal printer are compatible with any Ohaus balance with an RS232 interface. They are particularly useful when coupled to an Ohaus balance with GLP functionality as they can provide a hard copy audit trail. A printer cable is required (but not supplied) for specific Ohaus balances.
Ordering information: Der er et stort udvalg af tilbehør og reservedele til Ohaus vægte og de mest almindeligt anvendte fremgår nedenfor: For øvrige varer bedes du gå ind på det lokale Avantor websted.
Documents
Specifications
- Beskrivelse:Printer cable and adapter (STP103) for Navigator™ and Scout™ Pro (with RS232 option)
- Seq:22