Stands for MSC-Advantage
Manually adjustable floor stand, 750 to 950 mm working height in 50 mm increments, for 1,8 m cabinet
Catalog # 135-1317
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
Specifications
- Beskrivelse:Manually adjustable floor stand, 750 to 950 mm working height in 50 mm increments, for 1,8 m cabinet
- Seq:08