Orders LinkContactUsLinkComponent
Intravenøst kateter, Surflash™
Intravenøst kateter, G 20, lyserød, PU, 51 mm
Katalog # TERUSR-FF2051
Leverandør:   TERUMO
Intravenøst kateter, Surflash™
Katalog # TERUSR-FF2051
Leverandør:  TERUMO
Intravenøst kateter, Surflash™
Intravenøst kateter, G 20, lyserød, PU, 51 mm
Price
716,00 kr
Log ind for tilgængelighed
1 * 50 Items
Product Details & Documents
Surflash is made of PU, which is firm at room temperature but soft at body temperature, making the catheter more comfortable.
  • Only exists in the straight version
  • Enables quick and easy confirmation of the placement in the vein by an additional flashback
  • Higher certainty of proper placement
Helpful with patients whose veins are hard to detect and in which venipuncture is more difficult, such as small children, people undergoing chemotherapy.
Specifikationer
  • Måler:
    G 20
  • Farve:
    lyserød
  • Beskrivelse:
    Intravenøst kateter
  • Længde:
    51 mm
  • Materiale:
    PU
  • Seq:
    006